Mal-PEG4-Acid ≥97%

Supplier: Ambeed
Warning

Synonyms: 1-(2,5-Dioxo-2,5-dihydro-1H-pyrrol-1-yl)-3,6,9,12-tetraoxapentadecan-15-oic acid

A366905-100MG A366905-1G A366905-250MG
77616-602EA 191.58 USD
77616-602 77748-730 77748-731
Mal-PEG4-Acid ≥97%
Mal-PEG4-Acid

Precaución: for Research Use Only

Formula: C₁₅H₂₃NO₈
MW: 345.345 g/mol
MDL Number: MFCD18916986
CAS Number: 518044-41-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR