(1S)-1-Cyclopropyl-2,2,2-trifluoroethan-1-amine hydrochloride ≥95%
Supplier: Ambeed
|
Warning
|
A372172-1G
A372172-100MG
A372172-250MG
77572-114EA
283
USD
77572-114
77900-699
77901-537
(1S)-1-Cyclopropyl-2,2,2-trifluoroethan-1-amine hydrochloride ≥95%
(1S)-1-Cyclopropyl-2,2,2-trifluoroethan-1-amine hydrochloride
Precaución: for Research Use Only
Formula:
C₅H₈F₃N·ClH |
MDL Number:
MFCD26569801 CAS Number: 1338377-73-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...