2-Chloro-5-(hydroxymethyl)phenylboronic acid ≥98%

Supplier: Ambeed
Danger

Synonyms: 2-Chloro-5-hydroxymethylphenylboronic acid

A372130-5G A372130-250MG
77748-745EA 67 USD
77748-745 77748-744
2-Chloro-5-(hydroxymethyl)phenylboronic acid ≥98%
2-Chloro-5-(hydroxymethyl)phenylboronic acid

Precaución: for Research Use Only

Formula: C₇H₈BClO₃
MW: 186.4000 g/mol
MDL Number: MFCD06656270
CAS Number: 1003042-59-8
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR