Lanosterol 60%

Supplier: Ambeed
Warning

A371706-5G A371706-25G A371706-100G A371706-1G A371706-100MG
77572-110EA 22 USD
77572-110 77601-646 77668-730 77668-732 77879-810
Lanosterol 60%
Lanosterol

Precaución: for Research Use Only

Formula: C₃₀H₅₀O
MDL Number: MFCD00021108
CAS Number: 79-63-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR