7-Amino-4-methyl-3-coumarinylacetic acid (AMCA-H) ≥97%

Supplier: Ambeed
Warning

Synonyms: AMCA, 2H-1-Benzopyran-3-acetic acid, 7-Amino-4-methylcoumarin-3-acetic acid, Aminomethyl coumarin acetic acid, 2-(7-Amino-4-methyl-2-oxo-2H-chromen-3-yl)acetic acid

A371673-250MG A371673-100MG
77601-644EA 125.05 USD
77601-644 77668-728
7-Amino-4-methyl-3-coumarinylacetic acid (AMCA-H) ≥97%
7-Amino-4-methyl-3-coumarinylacetic acid (AMCA-H)

Precaución: for Research Use Only

Formula: C₁₂H₁₁NO₄
MW: 233.22 g/mol
MDL Number: MFCD00081051
CAS Number: 106562-32-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR