3-(4-Bromophenyl)acrylaldehyde ≥97%

Supplier: Ambeed
Warning

A371527-100G A371527-5G
77748-741EA 705 USD
77748-741 77748-742
3-(4-Bromophenyl)acrylaldehyde ≥97%
3-(4-Bromophenyl)acrylaldehyde

Precaución: for Research Use Only

Formula: C₉H₇BrO
MW: 211.0553 g/mol
MDL Number: MFCD03032509
CAS Number: 3893-18-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR