(1aR,1bR,4aS,7R,7aS,9aR)-7,9a-Dimethyl-4-methylenedecahydro-3H-oxireno[2',3':7,8]naphtho[8a,1-b]furan-3-one ≥98%

Supplier: Ambeed
Warning

Synonyms: Arteannuin B

A365685-5MG A365685-50MG
77626-708EA 54.59 USD
77626-708 77878-076
(1aR,1bR,4aS,7R,7aS,9aR)-7,9a-Dimethyl-4-methylenedecahydro-3H-oxireno[2',3':7,8]naphtho[8a,1-b]furan-3-one ≥98%
(1aR,1bR,4aS,7R,7aS,9aR)-7,9a-Dimethyl-4-methylenedecahydro-3H-oxireno[2',3':7,8]naphtho[8a,1-b]furan-3-one

Precaución: for Research Use Only

Formula: C₁₅H₂₀O₃
MW: 248.32 g/mol
MDL Number: MFCD09859140
CAS Number: 50906-56-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR