4,4',4''-Trimethyl-2,2':6',2''-terpyridine ≥98%, Ambeed.Inc

Supplier: Ambeed
Warning

A365186-100MG A365186-250MG A365186-1G
77613-082EA 64.89 USD
77613-082 77684-067 77890-750
4,4',4''-Trimethyl-2,2':6',2''-terpyridine ≥98%, Ambeed.Inc
4,4',4''-Trimethyl-2,2':6',2''-terpyridine

Precaución: For research use only.

Formula: C₁₈H₁₇N₃
MDL Number: MFCD06796994
CAS Number: 33354-75-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR