5-Bromo-2-(trifluoromethoxy)pyridine ≥98%, Ambeed.Inc
Supplier: Ambeed
|
Warning
|
A371040-1G
A371040-250MG
77684-075EA
468
USD
77684-075
77684-076
5-Bromo-2-(trifluoromethoxy)pyridine ≥98%, Ambeed.Inc
5-Bromo-2-(trifluoromethoxy)pyridine
Precaución: For research use only.
Formula:
C₆H₃BrF₃NO MW: 241.99 g/mol |
MDL Number:
MFCD07374891 CAS Number: 886371-77-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...