3,4-Dichloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine ≥98%

Supplier: Ambeed
Danger

A364902-250MG A364902-100MG A364902-1G
77746-727EA 187 USD
77746-727 77746-725 77746-726
3,4-Dichloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine ≥98%
3,4-Dichloro-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

Precaución: for Research Use Only

Formula: C₁₁H₁₄BCl₂NO₂
MW: 273.95 g/mol
MDL Number: MFCD18712632
CAS Number: 1451391-08-4
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR