2,2'-((2-Chloro-5-cyano-1,3-phenylene)bis(azanediyl))bis(2-oxoacetic acid) ≥98%

Supplier: Ambeed
Danger

Synonyms: Lodoxamide

A370864-10MG A370864-5MG
77668-722EA 53 USD
77668-722 77668-724
2,2'-((2-Chloro-5-cyano-1,3-phenylene)bis(azanediyl))bis(2-oxoacetic acid) ≥98%
2,2'-((2-Chloro-5-cyano-1,3-phenylene)bis(azanediyl))bis(2-oxoacetic acid)

Precaución: for Research Use Only

Formula: C₁₁H₆ClN₃O₆
MW: 311.63 g/mol
MDL Number: MFCD00864803
CAS Number: 53882-12-5
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR