Ibutamoren mesylate ≥98%

Supplier: Ambeed
Warning

Synonyms: L 163191, MK0677, L163191, L-163191, Ibutamoren, 2-amino-2-methyl-N-[(2R)-1-(1-methylsulphonylspiro[2H-indole-3,4'-piperidine]-1'-yl)-1-oxo-3-(phenylmethoxy)propan-2-yl]propanamide mesylate, MK-0677, (R)-2-Amino-N-(3-(benzyloxy)-1-(1-(methylsulphonyl)spiro[indoline-3,4'-piperidin]-1'-yl)-1-oxopropan-2-yl)-2-methylpropanamide methanesulphonate

A370432-10MG A370432-1MG A370432-5MG A370432-1G A370432-250MG
77594-100EA 9.23 USD
77594-100 77594-102 77594-104 77616-630 77616-632
Ibutamoren mesylate ≥98%
Ibutamoren mesylate

Precaución: for Research Use Only

Formula: C₂₇H₃₆N₄O₅S.CH₃SO₃H
MW: 624.77 g/mol
MDL Number: MFCD00942284
CAS Number: 159752-10-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR