Thapsigargin ≥99% (by HPLC)
Supplier: Ambeed
|
Danger
|
Synonyms:
(3S,3aR,4S,6S,6aR,7S,8S,9bS)-6-Acetoxy-4-(butyryloxy)-3,3a-dihydroxy-3,6,9-trimethyl-8-(((Z)-2-methylbut-2-enoyl)oxy)-2-oxo-2,3,3a,4,5,6,6a,7,8,9b-decahydroazuleno[4,5-b]furan-7-yl octanoate
A370143-100MG
A370143-10MG
A370143-1MG
A370143-50MG
A370143-5MG
77668-704EA
908
USD
77668-704
77668-706
77668-708
77668-710
77668-712
Thapsigargin ≥99% (by HPLC)
Thapsigargin
Precaución: for Research Use Only
Formula:
C₃₄H₅₀O₁₂ MW: 650.762 g/mol |
MDL Number:
MFCD00083511 CAS Number: 67526-95-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...