Oleuropein ≥98%

Supplier: Ambeed
Warning

Synonyms: Methyl (2S,4S,E)-4-(2-(3,4-dihydroxyphenethoxy)-2-oxoethyl)-3-ethylidene-2-(((2S,3R,4S,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)tetrahydro-2H-pyran-2-yl)oxy)-3,4-dihydro-2H-pyran-5-carboxylate

A369995-100MG A369995-250MG A369995-50MG
77672-846EA 83 USD
77672-846 77672-848 77672-850
Oleuropein ≥98%
Oleuropein

Precaución: for Research Use Only

Formula: C₂₅H₃₂O₁₃
MW: 540.51 g/mol
MDL Number: MFCD16879008
CAS Number: 32619-42-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR