1-Hydroxypyridine-2(1H)-thione ≥98%

Supplier: Ambeed
Warning

Synonyms: Pyrithione

A369732-100MG A369732-1G A369732-250MG
77613-094EA 104.03 USD
77613-094 77672-840 77672-842
1-Hydroxypyridine-2(1H)-thione ≥98%
1-Hydroxypyridine-2(1H)-thione

Precaución: for Research Use Only

Formula: C₅H₅NOS
MW: 127.16 g/mol
MDL Number: MFCD00059791
CAS Number: 1121-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR