4-Amino-3-methoxybenzotrifluoride ≥98%

Supplier: Ambeed
Warning

Synonyms: 2-Amino-5-(trifluoromethyl)anisole, 2-Methoxy-4-(trifluoromethyl)aniline

A363274-1G A363274-250MG A363274-100MG
77613-068EA 151.41 USD
77613-068 77613-070 77672-826
4-Amino-3-methoxybenzotrifluoride ≥98%
4-Amino-3-methoxybenzotrifluoride

Precaución: for Research Use Only

Formula: C₈H₈F₃NO
MW: 191.15 g/mol
MDL Number: MFCD04115973
CAS Number: 158727-56-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR