cis-3-Aminocyclohexanecarbonitrile hydrochloride 95+%

Supplier: Ambeed
Warning

A362969-100MG A362969-1G A362969-250MG A362969-5G
77686-027EA 94 USD
77686-027 77901-358 77900-761 77901-168
cis-3-Aminocyclohexanecarbonitrile hydrochloride 95+%
cis-3-Aminocyclohexanecarbonitrile hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₃ClN₂
MDL Number: MFCD22548414
CAS Number: 1403323-06-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR