Deoxycholic acid ≥98%

Supplier: Ambeed
Warning

A359830-100G A359830-100MG A359830-1G A359830-5G A359830-250MG A359830-25G A359830-500G
77601-592EA 79.95 USD
77601-592 77601-594 77601-596 77601-598 77879-772 77879-773 77879-774
Deoxycholic acid ≥98%
Deoxycholic acid

Precaución: for Research Use Only

Formula: C₂₄H₄₀O₄
MDL Number: MFCD00003673
CAS Number: 83-44-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR