(4-(1-(Trifluoromethyl)cyclopropyl)phenyl)boronic acid ≥95%

Supplier: Ambeed
Warning

A362753-100MG A362753-1G A362753-250MG
77890-739EA 340 USD
77890-739 77890-740 77890-741
(4-(1-(Trifluoromethyl)cyclopropyl)phenyl)boronic acid ≥95%
(4-(1-(Trifluoromethyl)cyclopropyl)phenyl)boronic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀BF₃O₂
MDL Number: MFCD26406755
CAS Number: 1431616-41-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR