Sb-9200 98+%

Supplier: Ambeed
Warning

Synonyms: Inarigivir soproxil, (((((2R,3S,5R)-5-(6-Amino-9H-purin-9-yl)-3-hydroxytetrahydrofuran-2-yl)methoxy)(((2R,3R,4R,5R)-5-(2,4-dioxo-3,4-dihydropyrimidin-1(2H)-yl)-2-(hydroxymethyl)-4-methoxytetrahydrofuran-3-yl)oxy)phosphoryl)thio)methyl isopropyl carbonate

A359450-5MG A359450-10MG
77748-716EA 83 USD
77748-716 77748-715
Sb-9200 98+%
Sb-9200

Precaución: for Research Use Only

Formula: C₂₅H₃₄N₇O₁₃PS
MDL Number: MFCD31715418
CAS Number: 942123-43-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR