(+)-Diacetyl-L-tartaric anhydride ≥97%

Supplier: Ambeed
Warning

Synonyms: (3R,4R)-2,5-Dioxotetrahydrofuran-3,4-diyl diacetate

A362177-25G A362177-5G A362177-100G
77616-586EA 31.93 USD
77616-586 77616-588 77686-025
(+)-Diacetyl-L-tartaric anhydride ≥97%
(+)-Diacetyl-L-tartaric anhydride

Precaución: for Research Use Only

Formula: C₈H₈O₇
MDL Number: MFCD00037918
CAS Number: 6283-74-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR