(4-Fluoro-2-methylphenyl)hydrazine hydrochloride ≥97%

Supplier: Ambeed
Warning

A361859-10G A361859-5G
77901-296EA 227 USD
77901-296 77900-747
(4-Fluoro-2-methylphenyl)hydrazine hydrochloride ≥97%
(4-Fluoro-2-methylphenyl)hydrazine hydrochloride

Precaución: for Research Use Only

Formula: C₇H₁₀ClFN₂
MDL Number: MFCD15209528
CAS Number: 439863-62-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR