1-Cyclopropylpyrazole-4-boronic Acid 98+%
Supplier: Ambeed
|
Warning
|
A361708-1G
A361708-250MG
77684-062EA
559
USD
77684-062
77684-063
1-Cyclopropylpyrazole-4-boronic Acid 98+%
1-Cyclopropylpyrazole-4-boronic Acid
Precaución: For research use only.
Formula:
C₆H₉BN₂O₂ MW: 151.96 g/mol |
MDL Number:
MFCD16621238 CAS Number: 1678534-30-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...