2-(1-Methyl-1H-pyrazol-4-yl)acetic acid ≥95%

Supplier: Ambeed
Warning

A358679-1G A358679-250MG
77881-977EA 291 USD
77881-977 77881-978
2-(1-Methyl-1H-pyrazol-4-yl)acetic acid ≥95%
2-(1-Methyl-1H-pyrazol-4-yl)acetic acid

Precaución: for Research Use Only

Formula: C₆H₈N₂O₂
MDL Number: MFCD09925217
CAS Number: 1152582-56-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR