Oligomycin A ≥99%

Supplier: Ambeed
Warning

Synonyms: (1R,2'R,4E,5'S,6S,6'S,7R,8S,10R,11R,12S,14R,15S,16R,18E,20E,22R,25S,28S,29R)-22-Ethyl-7,11,14,15-tetrahydroxy-6'-((R)-2-hydroxypropyl)-5',6,8,10,12,14,16,28,29-nonamethyl-3',4',5',6'-tetrahydro-2,26-dioxaspiro[bicyclo[23.3.1]nonacosa[4,18,20]triene-27,2'-pyran]-3,9,13-trione

A358360-1MG A358360-5MG
77594-064EA 60.48 USD
77594-064 77900-869
Oligomycin A ≥99%
Oligomycin A

Precaución: for Research Use Only

Formula: C₄₅H₇₄O₁₁
MDL Number: MFCD00065705
CAS Number: 579-13-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR