(1R,4S,5S,6S)-4-Amino-2-thiabicyclo[3.1.0]hexane-4,6-dicarboxylic acid 2,2-dioxide ≥98%

Supplier: Ambeed
Warning

Synonyms: LY404039

A358160-10MG A358160-5MG
77668-666EA 84 USD
77668-666 77668-668
(1R,4S,5S,6S)-4-Amino-2-thiabicyclo[3.1.0]hexane-4,6-dicarboxylic acid 2,2-dioxide ≥98%
(1R,4S,5S,6S)-4-Amino-2-thiabicyclo[3.1.0]hexane-4,6-dicarboxylic acid 2,2-dioxide

Precaución: for Research Use Only

Formula: C₇H₉NO₆S
MW: 235.21 g/mol
MDL Number: MFCD11974011
CAS Number: 635318-11-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR