Fmoc-O-t-butyl-L-β-homotyrosine ≥98%

Supplier: Ambeed
Warning

Synonyms: Fmoc-β-HoTyr(tBu)-OH, Fmoc-β-HomoTyr(tBu)-OH

A361394-5G A361394-100MG
77572-070EA 353.29 USD
77572-070 77672-816
Fmoc-O-t-butyl-L-β-homotyrosine ≥98%
Fmoc-O-t-butyl-L-beta-homotyrosine

Precaución: for Research Use Only

Formula: C₂₉H₃₁NO₅
MW: 473.57 g/mol
MDL Number: MFCD01862862
CAS Number: 219967-69-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR