4-Ethynylpyridin-2(1H)-one ≥95%

Supplier: Ambeed
Warning

A357666-100MG A357666-250MG A357666-50MG A357666-1G
77616-574EA 129.78 USD
77616-574 77748-709 77748-710 77748-708
4-Ethynylpyridin-2(1H)-one ≥95%
4-Ethynylpyridin-2(1H)-one

Precaución: for Research Use Only

Formula: C₇H₅NO
MDL Number: MFCD13189749
CAS Number: 1196152-08-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR