2-(1-(2-((3R,5S)-1-(3-Acetoxy-2,2-dimethylpropyl)-7-chloro-5-(2,3-dimethoxyphenyl)-2-oxo-1,2,3,5-tetrahydrobenzo[e][1,4]oxazepin-3-yl)acetyl)piperidin-4-yl)acetic acid ≥98%

Supplier: Ambeed
Warning

Synonyms: Lapaquistat acetate

A357554-5MG A357554-1MG
77613-032EA 203.94 USD
77613-032 77684-051
2-(1-(2-((3R,5S)-1-(3-Acetoxy-2,2-dimethylpropyl)-7-chloro-5-(2,3-dimethoxyphenyl)-2-oxo-1,2,3,5-tetrahydrobenzo[e][1,4]oxazepin-3-yl)acetyl)piperidin-4-yl)acetic acid ≥98%
2-(1-(2-((3R,5S)-1-(3-Acetoxy-2,2-dimethylpropyl)-7-chloro-5-(2,3-dimethoxyphenyl)-2-oxo-1,2,3,5-tetrahydrobenzo[e][1,4]oxazepin-3-yl)acetyl)piperidin-4-yl)acetic acid

Precaución: For research use only.

Formula: C₃₃H₄₁ClN₂O₉
MW: 645.14 g/mol
MDL Number: MFCD06407873
CAS Number: 189060-13-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR