3-Hydrazinobenzoic Acid Hydrochloride ≥98%
Supplier: Ambeed
|
Warning
|
A361048-25G
A361048-5G
77684-059EA
54
USD
77684-059
77684-060
3-Hydrazinobenzoic Acid Hydrochloride ≥98%
3-Hydrazinobenzoic Acid Hydrochloride
Precaución: For research use only.
Formula:
C₇H₉ClN₂O₂ MW: 188.611 g/mol |
MDL Number:
MFCD11226177 CAS Number: 87565-98-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...