N',N'''-(((1,4-Phenylenebis(methaneylylidene))bis(azaneylylidene))bis(4,1-phenylene))bis(N,N-dimethylacetimidamide) ≥98%

Supplier: Ambeed
Danger

A357500-10MG A357500-5MG
77748-706EA 73 USD
77748-706 77748-707
N',N'''-(((1,4-Phenylenebis(methaneylylidene))bis(azaneylylidene))bis(4,1-phenylene))bis(N,N-dimethylacetimidamide) ≥98%
N',N'''-(((1,4-Phenylenebis(methaneylylidene))bis(azaneylylidene))bis(4,1-phenylene))bis(N,N-dimethylacetimidamide)

Precaución: for Research Use Only

Formula: C₂₈H₃₂N₆
MDL Number: MFCD00872821
CAS Number: 115103-15-6
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR