2-Chloro-4-nitrobenzaldehyde ≥98%, Ambeed.Inc

Supplier: Ambeed
Warning

A352008-1G A352008-250MG
77684-040EA 167 USD
77684-040 77684-041
2-Chloro-4-nitrobenzaldehyde ≥98%, Ambeed.Inc
2-Chloro-4-nitrobenzaldehyde

Precaución: For research use only.

Formula: C₇H₄ClNO₃
MW: 185.565 g/mol
MDL Number: MFCD09842446
CAS Number: 5568-33-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR