Polyethylene glycol 800

Supplier: Ambeed
Warning

Synonyms: PEG800, Ethylene glycol polymer 800, PEG 800

A351909-1000G A351909-500G
77622-288EA 11.33 USD
77622-288 77622-290
Polyethylene glycol 800
Polyethylene glycol 800

Precaución: for Research Use Only

Formula: H(OCH₂CH₂)nOH
MDL Number: MFCD00081839
CAS Number: 25322-68-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR