BQ-788 sodium salt ≥98%

Supplier: Ambeed
Warning

Synonyms: ETB Receptor Antagonist, Sodium (R)-2-((R)-2-((S)-2-((2R,6S)-2,6-dimethylpiperidine-1-carboxamido)-4,4-dimethylpentanamido)-3-(1-(methoxycarbonyl)-1H-indol-3-yl)propanamido)hexanoate

A356845-1MG A356845-10MG A356845-5MG
77616-562EA 89.61 USD
77616-562 77746-701 77746-702
BQ-788 sodium salt ≥98%
BQ-788 sodium salt

Precaución: for Research Use Only

Formula: C₃₄H₅₀N₅O₇·Na
MDL Number: MFCD00797882
CAS Number: 156161-89-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR