(4-(tert-Butoxycarbonyl)-3,4-dihydro-2H-benzo[b][1,4]oxazin-7-yl)boronic acid ≥97%

Supplier: Ambeed
Warning

A351789-100MG A351789-250MG
77685-997EA 218 USD
77685-997 77685-998
(4-(tert-Butoxycarbonyl)-3,4-dihydro-2H-benzo[b][1,4]oxazin-7-yl)boronic acid ≥97%
(4-(tert-Butoxycarbonyl)-3,4-dihydro-2H-benzo[b][1,4]oxazin-7-yl)boronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₈BNO₅
MDL Number: MFCD28061758
CAS Number: 2304633-98-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR