2-Thioxo-5-((5-(3-(trifluoromethyl)phenyl)furan-2-yl)methylene)thiazolidin-4-one ≥95%
Supplier: Ambeed
|
Warning
|
Synonyms:
CCR-11
A356530-100MG
A356530-10MG
A356530-5MG
77686-012EA
453
USD
77686-012
77686-013
77686-014
2-Thioxo-5-((5-(3-(trifluoromethyl)phenyl)furan-2-yl)methylene)thiazolidin-4-one ≥95%
2-Thioxo-5-((5-(3-(trifluoromethyl)phenyl)furan-2-yl)methylene)thiazolidin-4-one
Precaución: for Research Use Only
Formula:
C₁₅H₈F₃NO₂S₂ MW: 355.35 g/mol |
MDL Number:
MFCD00640629 CAS Number: 301687-87-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...