Methanesulfonato[2-(di-t-butylphosphino)-2'-(N,N-dimethylamino)-1,1'-biphenyl](2'-amino-1,1'-biphenyl-2-yl)palladium(II) ≥98%

Supplier: Ambeed
Warning

A351464-250MG A351464-100MG A351464-1G
77616-542EA 47.38 USD
77616-542 77746-690 77746-691
Methanesulfonato[2-(di-t-butylphosphino)-2'-(N,N-dimethylamino)-1,1'-biphenyl](2'-amino-1,1'-biphenyl-2-yl)palladium(II) ≥98%
Methanesulfonato[2-(di-t-butylphosphino)-2'-(N,N-dimethylamino)-1,1'-biphenyl](2'-amino-1,1'-biphenyl-2-yl)palladium(II)

Precaución: for Research Use Only

Formula: C₃₅H₄₄N₂O₃PPdS
MW: 710.19 g/mol
MDL Number: MFCD29037178
CAS Number: 1445085-92-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR