Selumetinib ≥98%

Supplier: Ambeed
Warning

Synonyms: 5-((4-Bromo-2-chlorophenyl)amino)-4-fluoro-N-(2-hydroxyethoxy)-1-methyl-1H-benzo[d]imidazole-6-carboxamide, AZD6244

A351367-1MG A351367-250MG A351367-100MG A351367-50MG
77601-572EA 7.18 USD
77601-572 77601-574 77668-616 77668-618
Selumetinib ≥98%
Selumetinib

Precaución: for Research Use Only

Formula: C₁₇H₁₅BrClFN₄O₃
MW: 457.7 g/mol
MDL Number: MFCD11977472
CAS Number: 606143-52-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR