2-(4-Cyanophenyl)-2-methylpropanoic acid ≥95%, Ambeed.Inc

Supplier: Ambeed
Danger

A351250-1G A351250-100MG A351250-5G
77684-037EA 144 USD
77684-037 77892-722 77892-723
2-(4-Cyanophenyl)-2-methylpropanoic acid ≥95%, Ambeed.Inc
2-(4-Cyanophenyl)-2-methylpropanoic acid

Precaución: For research use only.

Formula: C₁₁H₁₁NO₂
MDL Number: MFCD19695824
CAS Number: 1000804-51-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR