4-Methyl-1H-1,2,3-benzotriazole ≥96%

Supplier: Ambeed
Warning

Synonyms: 4-Methyl-1H-benzo[d][1,2,3]triazole

A355668-250MG A355668-5G
77672-778EA 80 USD
77672-778 77872-539
4-Methyl-1H-1,2,3-benzotriazole ≥96%
4-Methyl-1H-1,2,3-benzotriazole

Precaución: for Research Use Only

Formula: C₇H₇N₃
MDL Number: MFCD00035800
CAS Number: 29878-31-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR