2-Iodo-DL-phenylalanine ≥95%

Supplier: Ambeed
Danger

A350799-1G A350799-250MG
77900-746EA 138 USD
77900-746 77901-188
2-Iodo-DL-phenylalanine ≥95%
2-Iodo-DL-phenylalanine

Precaución: for Research Use Only

Formula: C₉H₁₀INO₂
MDL Number: MFCD05227973
CAS Number: 1986-86-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR