2-(3-Acetylphenyl)acetic acid ≥95%

Supplier: Ambeed
Warning

A350728-100MG A350728-1G
77616-538EA 216.3 USD
77616-538 77616-540
2-(3-Acetylphenyl)acetic acid ≥95%
2-(3-Acetylphenyl)acetic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₃
MDL Number: MFCD16037305
CAS Number: 18749-47-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR