trans-Diamminedichloropalladium(II) 98+%

Supplier: Ambeed
Danger

Synonyms: trans-Dichlorodiammine palladium(II), Dichlorodiamminepalladium, trans-Diammine dichloropalladium

A350255-250MG A350255-100MG A350255-1G
77601-564EA 24.6 USD
77601-564 77668-610 77668-612
trans-Diamminedichloropalladium(II) 98+%
trans-Diamminedichloropalladium(II)

Precaución: for Research Use Only

Formula: Cl₂H₆N₂Pd
MW: 211.39 g/mol
MDL Number: MFCD00011621
CAS Number: 13782-33-7
UN: 2811
ADR: 6.1 Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR