Benzo[d]thiazole-7-carbaldehyde ≥97%
Supplier: Ambeed
|
Warning
|
A350001-100MG
A350001-50MG
77684-035EA
171
USD
77684-035
77684-036
Benzo[d]thiazole-7-carbaldehyde ≥97%
Benzo[d]thiazole-7-carbaldehyde
Precaución: For research use only.
Formula:
C₈H₅NOS MW: 163.2 g/mol |
MDL Number:
MFCD18822744 CAS Number: 144876-37-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...