7-Fluoroisatin ≥97%

Supplier: Ambeed
Warning

Synonyms: 7-Fluoro-1H-indole-2,3-dione, 7-Fluoroindoline-2,3-dione

A349806-100G A349806-5G A349806-1G
77572-022EA 188.49 USD
77572-022 77594-042 77601-560
7-Fluoroisatin ≥97%
7-Fluoroisatin

Precaución: for Research Use Only

Formula: C₈H₄FNO₂
MW: 165.12 g/mol
MDL Number: MFCD01569508
CAS Number: 317-20-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR