5-Nitro-2-phenylpyridine ≥98%

Supplier: Ambeed
Warning

A349669-1G A349669-250MG
77878-061EA 23 USD
77878-061 77878-062
5-Nitro-2-phenylpyridine ≥98%
5-Nitro-2-phenylpyridine

Precaución: for Research Use Only

Formula: C₁₁H₈N₂O₂
MDL Number: MFCD04114193
CAS Number: 89076-64-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR