Mesitylacetic acid ≥97%

Supplier: Ambeed
Warning

A349571-25G A349571-100G
77572-020EA 60 USD
77572-020 77668-598
Mesitylacetic acid ≥97%
Mesitylacetic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₄O₂
MDL Number: MFCD00014368
CAS Number: 4408-60-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR