KN-92 phosphate ≥98%

Supplier: Ambeed
Danger

Synonyms: KN92 phosphate, KN-92, phosphoric acid, KN 92 phosphate, KN-92 phosphate, N-[2-[[[(E)-3-(4-chlorophenyl)prop-2-enyl]-methylamino]methyl]phenyl]-4-methoxybenzenesulphonamide, KN92, N-(2-(((3-(4-Chlorophenyl)allyl)(methyl)amino)methyl)phenyl)-4-methoxybenzenesulphonamide phosphate

A349509-10MG A349509-1MG A349509-5MG A349509-100MG
77616-532EA 48.41 USD
77616-532 77616-534 77616-536 77685-992
KN-92 phosphate ≥98%
KN-92 phosphate

Precaución: for Research Use Only

Formula: C₂₄H₂₅ClN₂O₃S.H₃PO₄
MW: 554.98 g/mol
MDL Number: MFCD24369012
CAS Number: 1135280-28-2
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR