1-(2-Hydroxyethyl)-2-imidazolidinone ≥98%

Supplier: Ambeed
Warning

A349470-5G A349470-25G
77572-016EA 13 USD
77572-016 77746-686
1-(2-Hydroxyethyl)-2-imidazolidinone ≥98%
1-(2-Hydroxyethyl)-2-imidazolidinone

Precaución: for Research Use Only

Formula: C₅H₁₀N₂O₂
MDL Number: MFCD00037883
CAS Number: 3699-54-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR