2,6-Dimethyl-3,5-heptanedione ≥98%

Supplier: Ambeed
Danger

A349300-5G A349300-1G A349300-25G
77616-530EA 13.39 USD
77616-530 77746-683 77746-684
2,6-Dimethyl-3,5-heptanedione ≥98%
2,6-Dimethyl-3,5-heptanedione

Precaución: for Research Use Only

Formula: C₉H₁₆O₂
MDL Number: MFCD00015040
CAS Number: 18362-64-6
UN: 1224
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR